Assay Details
Target Gene Details
Entrez Gene ID: | 218333 |
Gene Name: | interactor of little elongation complex ELL subunit 1 |
Gene Aliases: |
C77245, mKIAA0947 |
Location: |
Chr.13:70588688-70637838 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ice1 | NM_144837.3 | NP_659086.2 | ||
| AK143610.1 | 3 | 247 |
More Information
Set Membership: |
|

Genomic Map