Assay Details
Target Gene Details
Entrez Gene ID: | 80837 |
Gene Name: | ras homolog gene family, member J |
Gene Aliases: |
1110005O19Rik, AW210585, Arhj, TC10L, TCL |
Location: |
Chr.12:75308313-75401455 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhoj | NM_023275.2 | NP_075764.1 | ||
| AB060651.1 | BAB91069.1 | |||
| AF309564.1 | AAL09441.1 | |||
| AJ276568.1 | CAC06700.1 | |||
| AK003482.1 | BAB22812.1 | |||
| AK003490.1 | BAB22818.1 | |||
| AK156619.1 | BAE33778.1 | |||
| AK157518.1 | BAE34111.1 | |||
| AK159385.1 | BAE35040.1 | |||
| AK166409.1 | ||||
| AK166480.1 | ||||
| AK181911.1 | ||||
| BC024626.1 | ||||
| BC043719.1 | AAH43719.1 | |||
| BC054464.1 | AAH54464.1 |
More Information
Set Membership: |
|

Genomic Map