Assay Details
Target Gene Details
Entrez Gene ID: | 13487 |
Gene Name: | solute carrier family 26, member 3 |
Gene Aliases: |
9030623B18Rik, 9130013M11Rik, AV376035, Dra |
Location: |
Chr.12:31390876-31473922 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc26a3 | NM_021353.3 | NP_067328.1 | ||
| AF136751.1 | AAD42784.1 | |||
| AK018566.1 | ||||
| AK035716.1 | ||||
| BC022639.1 | AAH22639.1 | |||
| BC037066.1 | AAH37066.1 | |||
| BC139271.1 | ||||
| BC139273.1 |
More Information
Set Membership: |
|

Genomic Map