Assay Details
Target Gene Details
Entrez Gene ID: | 212483 |
Gene Name: | family with sequence similarity 193, member B |
Gene Aliases: |
IRIZIO, Kiaa1931 |
Location: |
Chr.13:55539316-55571120 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam193b | NM_145382.4 | 10 | 3903 | NP_663357.2 |
| AK040568.1 | 4 | 2031 | BAC30629.1 | |
| AK155237.1 | 10 | 3897 | BAE33140.1 | |
| AK173303.1 | 10 | 2752 | BAD32581.1 | |
| BC021381.1 | 4 | 1493 | AAH21381.1 | |
| BC023015.1 | 4 | 1435 | AAH23015.1 | |
| BC025483.1 | 10 | 2762 | AAH25483.2 |

Genomic Map