Assay Details
Target Gene Details
Entrez Gene ID: | 19819 |
Gene Name: | ribonuclease H1 |
Gene Aliases: |
- |
Location: |
Chr.12:28649602-28659591 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnaseh1 | NM_001286865.1 | NP_001273794.1 | ||
| NM_011275.3 | NP_035405.2 | |||
| NR_104608.1 | ||||
| AF048993.1 | AAC78562.1 | |||
| AK011680.1 | ||||
| BC019411.1 | AAH19411.1 | |||
| BY274268.1 | ||||
| CA554498.1 | ||||
| CJ073105.1 |
More Information
Set Membership: |
|

Genomic Map