Assay Details
Target Gene Details
Entrez Gene ID: | 20338 |
Gene Name: | sel-1 suppressor of lin-12-like (C. elegans) |
Gene Aliases: |
AW493766, Sel1h, mKIAA4137 |
Location: |
Chr.12:91806043-91849157 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sel1l | NM_001039089.1 | 3 | 383 | NP_001034178.1 |
| NM_011344.2 | 3 | 383 | NP_035474.1 | |
| AF063095.1 | 3 | 332 | AAD05210.1 | |
| AK005023.2 | 3 | 356 | BAB23750.1 | |
| AK005544.1 | 3 | 366 | BAB24113.1 | |
| AK146022.1 | 3 | 357 | ||
| AK160420.1 | 3 | 365 | ||
| AK160936.1 | 3 | 341 | BAE36100.1 | |
| AK161546.1 | 3 | 357 | BAE36455.1 | |
| AK220502.1 | 3 | 383 | BAD90512.1 | |
| BC026816.1 | 3 | 355 | AAH26816.1 |

Genomic Map