Assay Details
Target Gene Details
Entrez Gene ID: | 23985 |
Gene Name: | solute carrier family 26, member 4 |
Gene Aliases: |
Pds, pendrin |
Location: |
Chr.12:31519814-31560637 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc26a4 | NM_011867.3 | 9 | 1295 | NP_035997.1 |
| AF167411.1 | 9 | 1296 | AAD51617.1 | |
| BC140995.1 | 8 | 1143 |
More Information
Set Membership: |
|

Genomic Map