Assay Details
Target Gene Details
Entrez Gene ID: | 320145 |
Gene Name: | trans-acting transcription factor 8 |
Gene Aliases: |
D930049B17Rik, mBtd |
Location: |
Chr.12:118845840-118852578 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sp8 | NM_177082.4 | 2 | 227 | NP_796056.2 |
| AK030745.1 | BAC27113.1 | |||
| AK053093.1 | 2 | 227 | BAC35264.1 | |
| BC082582.1 | AAH82582.1 |
More Information
Set Membership: |
|

Genomic Map