Assay Details
Target Gene Details
Entrez Gene ID: | 21340 |
Gene Name: | TATA box binding protein (Tbp)-associated factor, RNA polymerase I, B |
Gene Aliases: |
4930408G01Rik, A230108M10Rik, TAFI68, Tafi86, p63 |
Location: |
Chr.12:24498581-24558571 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf1b | NM_020614.2 | NP_065639.2 | ||
| NR_028341.1 | ||||
| AK015111.1 | 12 | 1278 | ||
| AK131963.1 | BAE20906.1 | |||
| AK165580.1 | BAE38271.1 | |||
| AK170313.1 | BAE41709.1 | |||
| BC057167.1 | ||||
| BC069907.1 | AAH69907.1 | |||
| BC094035.1 | AAH94035.1 | |||
| BC125027.1 | ||||
| BC125028.1 |

Genomic Map