Assay Details
Target Gene Details
Entrez Gene ID: | 74521 |
Gene Name: | protein phosphatase 4, regulatory subunit 4 |
Gene Aliases: |
8430415E04Rik, mKIAA1622 |
Location: |
Chr.12:103532289-103613839 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp4r4 | NM_028980.3 | 9 | 966 | NP_083256.2 |
| AK018414.1 | 8 | 725 | BAB31199.1 | |
| AK029499.1 | 9 | 967 | BAC26478.1 | |
| AK173221.1 | 7 | 684 | BAD32499.1 | |
| BC111887.1 | 9 | 961 | ||
| BC113754.1 | 9 | 961 |
More Information
Set Membership: |
|

Genomic Map