Assay Details
Target Gene Details
Entrez Gene ID: | 22018 |
Gene Name: | thyroid peroxidase |
Gene Aliases: |
- |
Location: |
Chr.12:30054661-30132984 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpo | NM_009417.2 | NP_033443.1 | ||
| AK047843.1 | BAC33171.1 | |||
| BC099421.1 | AAH99421.1 | |||
| X60703.1 | CAA43114.1 |
More Information
Set Membership: |
|

Genomic Map