Assay Details
Target Gene Details
Entrez Gene ID: | 238663 |
Gene Name: | spermatogenesis associated 31 subfamily D, member 1D |
Gene Aliases: |
4932411G14Rik, Fam75d1d |
Location: |
Chr.13:59725925-59731752 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata31d1d | NM_177711.3 | NP_808379.2 | ||
| AK077018.1 | BAC36569.1 |

Genomic Map