Assay Details
Target Gene Details
Entrez Gene ID: | 263406 |
Gene Name: | pleckstrin homology domain containing, family G (with RhoGef domain) member 3 |
Gene Aliases: |
BC030417 |
Location: |
Chr.12:76533247-76579042 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekhg3 | NM_153804.4 | NP_722499.4 | ||
| AK080928.1 | BAC38083.1 | |||
| AK162025.1 | BAE36687.1 | |||
| BC030417.1 | AAH30417.1 | |||
| BC049264.1 | AAH49264.1 | |||
| BC096443.1 | AAH96443.1 |

Genomic Map