Assay Details
Target Gene Details
Entrez Gene ID: | 80913 |
Gene Name: | pumilio RNA-binding family member 2 |
Gene Aliases: |
5730503J23Rik, Pumm2, mKIAA0235 |
Location: |
Chr.12:8674134-8752583 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pum2 | NM_001160219.1 | NP_001153691.1 | ||
| NM_001160220.1 | NP_001153692.1 | |||
| NM_001160221.1 | NP_001153693.1 | |||
| NM_001160222.1 | NP_001153694.1 | |||
| NM_030723.2 | NP_109648.2 | |||
| NR_027670.1 | ||||
| AF315590.1 | AAG31805.1 | |||
| AK048119.1 | ||||
| AK122225.1 | BAC65507.1 | |||
| AK134185.1 | ||||
| AK141682.1 | BAE24797.1 | |||
| AK147617.1 | ||||
| AK161357.1 | BAE36346.1 | |||
| AK163964.1 | BAE37556.1 | |||
| AY027917.1 | AAK21966.1 | |||
| BC041773.1 | AAH41773.1 |
More Information
Set Membership: |
|

Genomic Map