Assay Details
Target Gene Details
Entrez Gene ID: | 19878 |
Gene Name: | Rho-associated coiled-coil containing protein kinase 2 |
Gene Aliases: |
B230113H15Rik, ROKalpha, Rho-kinase, Rock-II, Rock2m, mKIAA0619 |
Location: |
Chr.12:16894978-16988274 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 22 - Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rock2 | NM_009072.2 | NP_033098.2 | ||
| AK035509.1 | BAC29084.1 | |||
| AK045517.1 | BAC32403.1 | |||
| AK172983.1 | BAD32261.1 | |||
| DQ864977.1 | ||||
| U58513.1 | AAC53133.1 |
More Information
Set Membership: |
|

Genomic Map