Assay Details
Target Gene Details
Entrez Gene ID: | 328133 |
Gene Name: | solute carrier family 39 (zinc transporter), member 9 |
Gene Aliases: |
2010002A02Rik, 2610511I23Rik, 4833420E20Rik, AW209151 |
Location: |
Chr.12:80644215-80683342 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc39a9 | NM_026244.2 | NP_080520.2 | ||
| AK014732.1 | ||||
| AK014734.1 | ||||
| AK080462.1 | BAC37924.1 | |||
| AK090062.1 | BAC41073.1 | |||
| BC117754.1 | ||||
| BC117755.1 | ||||
| BC158056.1 |
More Information
Set Membership: |
|

Genomic Map