Assay Details
Target Gene Details
Entrez Gene ID: | 192136 |
Gene Name: | succinyl-CoA glutarate-CoA transferase |
Gene Aliases: |
5033411D12Rik, D17907 |
Location: |
Chr.13:16857472-17694793 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sugct | NM_138654.3 | NP_619595.3 | ||
| AF397014.1 | AAL76419.1 | |||
| AK160265.1 | BAE35721.1 | |||
| BC055852.1 | AAH55852.1 | |||
| CJ068463.1 |
More Information
Set Membership: |
|

Genomic Map