Assay Details
Target Gene Details
Entrez Gene ID: | 19088 |
Gene Name: | protein kinase, cAMP dependent regulatory, type II beta |
Gene Aliases: |
AI451071, AW061005, PKARIIbeta, Pkarb2, RII(beta) |
Location: |
Chr.12:31958450-32061343 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkar2b | NM_011158.3 | NP_035288.2 | ||
| AK011647.1 | ||||
| AK041013.1 | BAC30779.1 | |||
| AK081805.1 | BAE43376.1 | |||
| AK135621.1 | ||||
| AK138963.1 | BAE23838.1 | |||
| BC048710.1 | AAH48710.1 |
More Information
Set Membership: |
|

Genomic Map