Assay Details
Target Gene Details
Entrez Gene ID: | 71907 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 9 |
Gene Aliases: |
2310014L03Rik, AW049635, Centerin |
Location: |
Chr.12:103996613-104013776 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpina9 | NM_027997.2 | NP_082273.1 | ||
| AK009343.1 | BAB26230.1 | |||
| BC132287.1 | ||||
| BC144769.1 |
More Information
Set Membership: |
|

Genomic Map