Assay Details
Target Gene Details
Entrez Gene ID: | 67241 |
Gene Name: | structural maintenance of chromosomes 6 |
Gene Aliases: |
2810489L22Rik, 3830418C19Rik, AA990493, AU018782, AW742439, SMC-6, Smc6l1, mKIAA4103 |
Location: |
Chr.12:11265886-11319785 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smc6 | NM_025695.4 | 17 | 2353 | NP_079971.2 |
| AJ310552.1 | 16 | 1945 | CAC39250.1 | |
| AK084304.1 | 3 | 3286 | ||
| AK135873.1 | 17 | 2091 | BAE22709.1 | |
| AK148478.1 | 17 | 2118 | BAE28576.1 | |
| AK220488.1 | 16 | 2028 | BAD90287.1 | |
| BC090630.1 | 17 | 2353 | AAH90630.1 | |
| BC099955.1 | 17 | 2040 | AAH99955.1 |
More Information
Set Membership: |
|

Genomic Map