Assay Details
Target Gene Details
Entrez Gene ID: | 94090 |
Gene Name: | tripartite motif-containing 9 |
Gene Aliases: |
- |
Location: |
Chr.12:70244533-70347761 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim9 | NM_001110202.1 | NP_001103672.1 | ||
| NM_001286387.1 | NP_001273316.1 | |||
| NM_001286388.1 | NP_001273317.1 | |||
| NM_053167.3 | NP_444397.2 | |||
| AK028280.1 | BAC25854.1 | |||
| AK049907.1 | 13 | 2483 | BAC33982.1 | |
| AK129109.1 | BAC97919.1 | |||
| AK163123.1 | BAE37203.1 |
More Information
Set Membership: |
|

Genomic Map