Assay Details
Target Gene Details
Entrez Gene ID: | 22630 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide |
Gene Aliases: |
2700028P07Rik, AA409740, AU021156, R74690 |
Location: |
Chr.12:21390329-21417436 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ywhaq | NM_011739.3 | NP_035869.1 | ||
| AK008948.1 | ||||
| AK012300.1 | ||||
| AK012540.1 | ||||
| AK145568.1 | BAE26517.1 | |||
| AK146371.1 | BAE27120.1 | |||
| AK150850.1 | BAE29907.1 | |||
| AK151437.1 | BAE30400.1 | |||
| AK151715.1 | BAE30634.1 | |||
| AK151864.1 | BAE30752.1 | |||
| AK159403.1 | BAE35055.1 | |||
| AK159817.1 | BAE35397.1 | |||
| AK163237.1 | BAE37249.1 | |||
| AK167397.1 | BAE39486.1 | |||
| AK200956.1 | ||||
| AK206650.1 | ||||
| BC080802.1 | AAH80802.1 | |||
| BC085299.1 | AAH85299.1 | |||
| BC090838.1 | AAH90838.1 | |||
| BC106164.1 | AAI06165.1 | |||
| BC132252.1 | ||||
| BC132530.1 | ||||
| D87662.1 | BAA13423.1 | |||
| U56243.1 | AAB72023.1 | |||
| U57312.1 | AAC53257.1 |
More Information
Set Membership: |
|

Genomic Map