Assay Details
Target Gene Details
Entrez Gene ID: | 72219 |
Gene Name: | spermatogenesis associated 31 subfamily D, member 1A |
Gene Aliases: |
1700013B16Rik, Fam75d1a, Fam75d3 |
Location: |
Chr.13:59700083-59706589 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata31d1a | NM_028157.2 | NP_082433.2 | ||
| AK005940.1 | BAB24327.1 | |||
| AK029673.1 | BAC26557.1 |
More Information
Set Membership: |
|

Genomic Map