Assay Details
Target Gene Details
Entrez Gene ID: | 105387 |
Gene Name: | aldo-keto reductase family 1, member C14 |
Gene Aliases: |
9030611N15Rik, AW557061 |
Location: |
Chr.13:4048999-4090560 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1c14 | NM_134072.1 | NP_598833.1 | ||
| AK033537.1 | BAC28346.1 | |||
| AK136528.1 | BAE23028.1 | |||
| AY730283.1 | AAU29338.1 | |||
| BC013482.1 | AAH13482.1 | |||
| BC026628.1 | AAH26628.1 |
More Information
Set Membership: |
|

Genomic Map