Assay Details
Target Gene Details
Entrez Gene ID: | 105349 |
Gene Name: | aldo-keto reductase family 1, member C18 |
Gene Aliases: |
AW146047 |
Location: |
Chr.13:4132615-4150651 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1c18 | NM_134066.2 | 2 | 169 | NP_598827.1 |
| AB059565.1 | 2 | 169 | BAB40958.1 | |
| AK153898.1 | 2 | 187 | BAE32242.1 | |
| BC034259.1 | 2 | 166 | AAH34259.1 |
More Information
Set Membership: |
|

Genomic Map