Assay Details
Target Gene Details
Entrez Gene ID: | 20135 |
Gene Name: | ribonucleotide reductase M2 |
Gene Aliases: |
AA407299, R2 |
Location: |
Chr.12:24708254-24714146 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrm2 | NM_009104.2 | NP_033130.1 | ||
| AK088907.1 | BAC40647.1 | |||
| AK142027.1 | BAE24918.1 | |||
| AK147111.1 | BAE27683.1 | |||
| AK161643.1 | BAE36508.1 | |||
| AK167078.1 | BAE39238.1 | |||
| AK167204.1 | BAE39331.1 | |||
| AK168117.1 | BAE40088.1 | |||
| AK168205.1 | BAE40165.1 | |||
| AK168994.1 | BAE40793.1 | |||
| AK188716.1 | ||||
| AK189805.1 | ||||
| BC085136.1 | AAH85136.1 | |||
| M14223.1 | AAA40062.1 |
More Information
Set Membership: |
|

Genomic Map