Assay Details
Target Gene Details
Entrez Gene ID: | 67241 |
Gene Name: | structural maintenance of chromosomes 6 |
Gene Aliases: |
2810489L22Rik, 3830418C19Rik, AA990493, AU018782, AW742439, SMC-6, Smc6l1, mKIAA4103 |
Location: |
Chr.12:11265886-11319785 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smc6 | NM_025695.4 | 25 | 3413 | NP_079971.2 |
| AJ310552.1 | 24 | 3005 | CAC39250.1 | |
| AK003872.1 | 4 | 366 | BAB23051.1 | |
| AK014442.1 | 6 | 617 | BAB29353.2 | |
| AK220488.1 | 24 | 3088 | BAD90287.1 | |
| BC026429.1 | 7 | 853 | AAH26429.1 | |
| BC090630.1 | 25 | 3413 | AAH90630.1 |
More Information
Set Membership: |
|

Genomic Map