Assay Details
Target Gene Details
Entrez Gene ID: | 432720 |
Gene Name: | aldo-keto reductase family 1, member C19 |
Gene Aliases: |
1810010N06Rik |
Location: |
Chr.13:4233504-4248361 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1c19 | NM_001013785.3 | NP_001013807.2 | ||
| BC087964.1 | AAH87964.1 | |||
| BG062632.2 |
More Information
Set Membership: |
|

Genomic Map