Assay Details
Target Gene Details
Entrez Gene ID: | 110095 |
Gene Name: | liver glycogen phosphorylase |
Gene Aliases: |
- |
Location: |
Chr.12:70190815-70227683 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pygl | NM_133198.2 | NP_573461.2 | ||
| AF288783.1 | AAG00588.1 | |||
| AK140321.1 | BAE24332.1 | |||
| AK145989.1 | BAE26811.1 | |||
| AK167337.1 | BAE39438.1 | |||
| BC013636.1 | AAH13636.1 |
More Information
Set Membership: |
|

Genomic Map