Assay Details
Target Gene Details
Entrez Gene ID: | 19325 |
Gene Name: | RAB10, member RAS oncogene family |
Gene Aliases: |
AW107754 |
Location: |
Chr.12:3247428-3309969 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab10 | NM_016676.5 | NP_057885.1 | ||
| AB232604.1 | BAF02866.1 | |||
| AF035646.1 | AAC29313.1 | |||
| AK008725.1 | BAB25858.1 | |||
| AK028320.1 | BAC25878.1 | |||
| AK087964.1 | BAC40062.1 | |||
| AK134988.1 | BAE22371.1 | |||
| AK146357.1 | BAE27109.1 | |||
| AK150319.1 | BAE29464.1 | |||
| AK150384.1 | BAE29515.1 | |||
| AK150791.1 | BAE29855.1 | |||
| AK151473.1 | BAE30429.1 | |||
| AK153315.1 | BAE31896.1 | |||
| AK153325.1 | BAE31904.1 | |||
| AK159383.1 | BAE35038.1 | |||
| AK159845.1 | BAE35423.1 | |||
| AK159988.1 | BAE35538.1 | |||
| AK160960.1 | BAE36116.1 | |||
| AK166010.1 | ||||
| AK168320.1 | BAE40259.1 | |||
| AK190935.1 | ||||
| AK193085.1 | ||||
| AK195457.1 | ||||
| AK202554.1 | ||||
| AK212462.1 | ||||
| BC056374.1 | AAH56374.1 | |||
| CT010394.1 | CAJ18601.1 |

Genomic Map