Assay Details
Target Gene Details
Entrez Gene ID: | 19325 |
Gene Name: | RAB10, member RAS oncogene family |
Gene Aliases: |
AW107754 |
Location: |
Chr.12:3247428-3309969 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab10 | NM_016676.5 | 5 | 1026 | NP_057885.1 |
| AB232604.1 | 5 | 475 | BAF02866.1 | |
| AF035646.1 | 5 | 716 | AAC29313.1 | |
| AK008725.1 | 5 | 1016 | BAB25858.1 | |
| AK028320.1 | 5 | 997 | BAC25878.1 | |
| AK087964.1 | 5 | 988 | BAC40062.1 | |
| AK134988.1 | 5 | 946 | BAE22371.1 | |
| AK146357.1 | 5 | 1027 | BAE27109.1 | |
| AK150319.1 | 5 | 986 | BAE29464.1 | |
| AK150384.1 | 5 | 680 | BAE29515.1 | |
| AK150791.1 | 5 | 976 | BAE29855.1 | |
| AK151473.1 | 5 | 680 | BAE30429.1 | |
| AK153315.1 | 5 | 986 | BAE31896.1 | |
| AK153325.1 | 5 | 976 | BAE31904.1 | |
| AK159383.1 | 5 | 991 | BAE35038.1 | |
| AK159845.1 | 5 | 991 | BAE35423.1 | |
| AK159988.1 | 5 | 985 | BAE35538.1 | |
| AK160960.1 | 5 | 981 | BAE36116.1 | |
| AK166010.1 | 5 | 1000 | ||
| AK168320.1 | 5 | 986 | BAE40259.1 | |
| AK190935.1 | 4 | 328 | ||
| AK193085.1 | 4 | 309 | ||
| AK202554.1 | 3 | 319 | ||
| AK212462.1 | 3 | 260 | ||
| BC056374.1 | 5 | 977 | AAH56374.1 | |
| CT010394.1 | 5 | 475 | CAJ18601.1 |

Genomic Map