Assay Details
Target Gene Details
Entrez Gene ID: | 72061 |
Gene Name: | RIKEN cDNA 2010111I01 gene |
Gene Aliases: |
2300006M17Rik, AP-O, Aopep, mir-23b, mir-24-1, mir-27b |
Location: |
Chr.13:62964893-63326096 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2010111I01Rik | NM_001289924.1 | 14 | 2450 | NP_001276853.1 |
| NM_001289926.1 | 14 | 2450 | NP_001276855.1 | |
| NR_110520.1 | 3 | 257 | ||
| NR_110521.1 | 3 | 257 | ||
| AJ810420.1 | 13 | 2164 | CAH17902.1 | |
| AK076298.1 | 3 | 213 | BAC36293.1 | |
| AK086954.1 | 3 | 288 | ||
| AK141931.1 | 13 | 2416 | BAE24891.1 | |
| AK167772.1 | 3 | 245 | BAE39804.1 | |
| BC006061.1 | 3 | 190 | ||
| BC048855.1 | 2 | 194 | AAH48855.1 | |
| BC064132.1 | 3 | 183 | AAH64132.1 | |
| BC065422.1 | 3 | 184 | AAH65422.1 | |
| BC141279.1 | 13 | 2301 |
More Information
Set Membership: |
|

Genomic Map