Assay Details
Target Gene Details
Entrez Gene ID: | 72061 |
Gene Name: | RIKEN cDNA 2010111I01 gene |
Gene Aliases: |
2300006M17Rik, AP-O, Aopep, mir-23b, mir-24-1, mir-27b |
Location: |
Chr.13:62964893-63326096 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2010111I01Rik | NM_001289924.1 | NP_001276853.1 | ||
| NM_001289926.1 | NP_001276855.1 | |||
| AJ810420.1 | CAH17902.1 | |||
| AK008397.1 | ||||
| AK141931.1 | BAE24891.1 | |||
| BC141279.1 |
More Information
Set Membership: |
|

Genomic Map