Assay Details
Target Gene Details
Entrez Gene ID: | 77337 |
Gene Name: | aldo-keto reductase family 1, member C21 |
Gene Aliases: |
9430025F20Rik, AI315367 |
Location: |
Chr.13:4574075-4586543 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1c21 | NM_029901.2 | NP_084177.2 | ||
| AB178898.1 | BAD18929.1 | |||
| AI035317.1 | ||||
| AK020439.1 | BAB32101.1 | |||
| AY742217.1 | AAW65159.1 | |||
| BC013531.1 | AAH13531.1 | |||
| BC061057.1 | AAH61057.1 | |||
| BC091761.1 | AAH91761.1 |
More Information
Set Membership: |
|

Genomic Map