Assay Details
Target Gene Details
Entrez Gene ID: | 77337 |
Gene Name: | aldo-keto reductase family 1, member C21 |
Gene Aliases: |
9430025F20Rik, AI315367 |
Location: |
Chr.13:4574075-4586543 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1c21 | NM_029901.2 | 9 | 1106 | NP_084177.2 |
| AI035317.1 | 1 | 117 | ||
| AK020439.1 | 9 | 1106 | BAB32101.1 | |
| BC013531.1 | 9 | 1082 | AAH13531.1 | |
| BC061057.1 | 9 | 1102 | AAH61057.1 | |
| BC091761.1 | 9 | 1082 | AAH91761.1 |
More Information
Set Membership: |
|

Genomic Map