Assay Details
Target Gene Details
Entrez Gene ID: | 56795 |
Gene Name: | ADP-ribosylation factor-like 10 |
Gene Aliases: |
Arl10a, Arm1 |
Location: |
Chr.13:54575000-54582765 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arl10 | NM_019968.2 | 3 | 506 | NP_064352.2 |
| AF205936.1 | 3 | 454 | AAF20926.1 | |
| AK154684.1 | 1 | 3790 | ||
| AK205970.1 | ||||
| BB857910.1 | ||||
| BC115498.2 | 3 | 447 |

Genomic Map