Assay Details
Target Gene Details
Entrez Gene ID: | 105171 |
Gene Name: | arrestin domain containing 3 |
Gene Aliases: |
AI450344, mKIAA1376 |
Location: |
Chr.13:80883338-80896042 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arrdc3 | NM_001042591.1 | NP_001036056.1 | ||
| AK084951.1 | BAC39320.1 | |||
| AK088288.1 | BAC40259.1 | |||
| AK122499.1 | BAC65781.1 | |||
| AK163364.1 | ||||
| BC031125.1 | AAH31125.1 | |||
| BC054844.1 | AAH54844.1 | |||
| BC098474.1 | AAH98474.1 | |||
| DQ861994.1 |

Genomic Map