Assay Details
Target Gene Details
Entrez Gene ID: | 11881 |
Gene Name: | arylsulfatase B |
Gene Aliases: |
1110007C02Rik, AI480648, As-1, As-1r, As-1s, As-1t, As1, As1-r, As1-s, As1-t, Asr-1, Ast-1 |
Location: |
Chr.13:93771023-93943016 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arsb | NM_009712.3 | 3 | 651 | NP_033842.3 |
| AK083309.1 | 3 | 635 | BAC38859.1 | |
| AK140163.1 | 3 | 701 | ||
| AK154098.1 | 3 | 571 | BAE32375.1 | |
| AK158312.1 | 3 | 652 | BAE34455.1 | |
| BC150661.1 | 3 | 608 | ||
| X92096.1 | 2 | 144 | CAA63067.1 |

Genomic Map