Assay Details
Target Gene Details
Entrez Gene ID: | 66603 |
Gene Name: | gem (nuclear organelle) associated protein 2 |
Gene Aliases: |
1700012N19Rik, Sip1 |
Location: |
Chr.12:59013393-59027988 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gemin2 | NM_025656.4 | NP_079932.2 | ||
| AK005928.1 | BAB24321.1 | |||
| AK007515.1 | BAB25083.1 | |||
| AK013414.1 | BAB28842.1 | |||
| BC053424.1 | AAH53424.1 |
More Information
Set Membership: |
|

Genomic Map