Assay Details
Target Gene Details
Entrez Gene ID: | 83702 |
Gene Name: | aldo-keto reductase family 1, member C6 |
Gene Aliases: |
3alpha-HSD, Akr1c1, Hsd17b5 |
Location: |
Chr.13:4434311-4457530 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akr1c6 | NM_030611.3 | NP_085114.1 | ||
| AK149458.1 | BAE28891.1 | |||
| BC056643.1 | AAH56643.1 | |||
| D45850.1 | BAA08285.1 |
More Information
Set Membership: |
|

Genomic Map