Assay Details
Target Gene Details
Entrez Gene ID: | 26898 |
Gene Name: | cathepsin J |
Gene Aliases: |
CATP, CatRLP, Ctsp |
Location: |
Chr.13:61000277-61005958 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsj | NM_012007.1 | 2 | 144 | NP_036137.1 |
| AF136272.1 | 2 | 144 | AAF13142.1 | |
| AF158182.1 | 1 | 81 | AAD41898.1 | |
| AK005526.1 | 2 | 148 | BAB24099.1 | |
| AK131661.1 | 2 | 148 | BAE20748.1 | |
| BC103769.1 | 2 | 137 | AAI03770.1 |
More Information
Set Membership: |
|

Genomic Map