Assay Details
Target Gene Details
Entrez Gene ID: | 218581 |
Gene Name: | DEP domain containing 1B |
Gene Aliases: |
9830132O03, AW260467, XTP1 |
Location: |
Chr.13:108316014-108389568 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Depdc1b | NM_178683.4 | 3 | 467 | NP_848798.1 |
| AK036550.1 | 3 | 426 | BAC29473.1 | |
| AK077676.1 | 3 | 468 | BAC36949.1 | |
| AK083480.1 | 3 | 463 | BAC38932.1 | |
| BC152330.1 | 3 | 472 | ||
| BC152331.1 | 3 | 472 |
More Information
Set Membership: |
|

Genomic Map