Assay Details
Target Gene Details
Entrez Gene ID: | 74145 |
Gene Name: | coagulation factor XIII, A1 subunit |
Gene Aliases: |
1200014I03Rik, AI462306, F13a |
Location: |
Chr.13:36867178-37050244 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F13a1 | NM_001166391.1 | 11 | 1453 | NP_001159863.1 |
| NM_028784.3 | 11 | 1478 | NP_083060.2 | |
| AK030914.1 | 12 | 1453 | BAE20463.1 | |
| AK036403.1 | 11 | 1479 | BAC29414.1 | |
| AK049092.1 | 11 | 1479 | BAC33539.1 | |
| AK165311.1 | 11 | 1479 | BAE38130.1 | |
| BC040274.1 | 11 | 1469 | AAH40274.1 |
More Information
Set Membership: |
|

Genomic Map