Assay Details
Target Gene Details
Entrez Gene ID: | 74145 |
Gene Name: | coagulation factor XIII, A1 subunit |
Gene Aliases: |
1200014I03Rik, AI462306, F13a |
Location: |
Chr.13:36867178-37050244 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F13a1 | NM_001166391.1 | 3 | 297 | NP_001159863.1 |
| NM_028784.3 | 3 | 322 | NP_083060.2 | |
| AK030914.1 | 3 | 297 | BAE20463.1 | |
| AK036403.1 | 3 | 323 | BAC29414.1 | |
| AK049092.1 | 3 | 323 | BAC33539.1 | |
| AK163591.1 | 3 | 297 | BAE37411.1 | |
| AK165311.1 | 3 | 323 | BAE38130.1 | |
| AK170215.1 | 3 | 322 | BAE41641.1 | |
| AK170278.1 | 3 | 323 | ||
| AK170860.1 | 3 | 323 | BAE42078.1 | |
| AK171273.1 | 3 | 323 | BAE42358.1 | |
| BC040274.1 | 3 | 313 | AAH40274.1 |
More Information
Set Membership: |
|

Genomic Map