Assay Details
Target Gene Details
Entrez Gene ID: | 16323 |
Gene Name: | inhibin beta-A |
Gene Aliases: |
- |
Location: |
Chr.13:16007745-16027723 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inhba | AK134991.1 | BAE22374.1 |

Genomic Map