Assay Details
Target Gene Details
Entrez Gene ID: | 17434 |
Gene Name: | molybdenum cofactor synthesis 2 |
Gene Aliases: |
AI415403 |
Location: |
Chr.13:114818237-114829420 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mocs2 | NM_001113374.1 | 1 | 38 | NP_001106845.1 |
| NM_001113375.1 | 1 | 38 | NP_001106846.1 | |
| NM_013826.3 | 1 | 38 | NP_038854.2 | |
| AF091872.1 | AAD14600.1 | |||
| AK002719.1 | BAB22306.1 | |||
| AK010112.1 | BAB26707.2 | |||
| AK078782.1 | BAC37393.1 | |||
| BC024371.1 | AAH24371.1 | |||
| CX220128.1 |
More Information
Set Membership: |
|

Genomic Map