Assay Details
Target Gene Details
Entrez Gene ID: | 218490 |
Gene Name: | basic transcription factor 3 |
Gene Aliases: |
1700054E11Rik |
Location: |
Chr.13:98309897-98317006 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btf3 | NM_001170540.1 | NP_001164011.1 | ||
| NM_145455.3 | NP_663430.2 | |||
| AK139664.1 | BAE24097.1 | |||
| AK146332.1 | BAE27087.1 | |||
| AK147171.1 | BAE27735.1 | |||
| AK160227.1 | ||||
| BC008233.1 | AAH08233.1 | |||
| BC064010.1 | AAH64010.1 | |||
| BC080837.1 | AAH80837.1 | |||
| CJ073405.1 |
More Information
Set Membership: |
|

Genomic Map