Assay Details
Target Gene Details
Entrez Gene ID: | 12231 |
Gene Name: | butyrophilin, subfamily 1, member A1 |
Gene Aliases: |
Btn |
Location: |
Chr.13:23456984-23465914 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btn1a1 | NM_013483.3 | NP_038511.1 | ||
| AK142596.1 | BAE25122.1 | |||
| AK145168.1 | BAE26272.1 | |||
| BC011497.1 | AAH11497.1 | |||
| BC031459.1 | AAH31459.1 | |||
| S80642.1 | AAB35893.1 |
More Information
Set Membership: |
|

Genomic Map