Assay Details
Target Gene Details
Entrez Gene ID: | 26898 |
Gene Name: | cathepsin J |
Gene Aliases: |
CATP, CatRLP, Ctsp |
Location: |
Chr.13:61000277-61005958 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctsj | NM_012007.1 | NP_036137.1 | ||
| AF136272.1 | AAF13142.1 | |||
| AF158182.1 | AAD41898.1 | |||
| AK005526.1 | BAB24099.1 | |||
| AK131661.1 | BAE20748.1 | |||
| BC103769.1 | AAI03770.1 |
More Information
Set Membership: |
|

Genomic Map