Assay Details
Target Gene Details
Entrez Gene ID: | 76577 |
Gene Name: | Fas associated factor family member 2 |
Gene Aliases: |
2210404D11Rik, AI462440, Ubxd8, mKIAA0887 |
Location: |
Chr.13:54621775-54664063 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Faf2 | NM_178397.3 | NP_848484.2 | ||
| AK122394.1 | BAC65676.1 | |||
| AK148529.1 | ||||
| AK151617.1 | BAE30554.1 | |||
| AK170111.1 | BAE41569.1 | |||
| BC046817.1 | AAH46817.1 | |||
| BY139990.1 |
More Information
Set Membership: |
|

Genomic Map